missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid, Glacial, Reagent, ACS, 99.7%, Spectrum™ Chemical

A1010, 64-19-7, C2H4O2

Supplier:  Spectrum Chemical Mfg Cor A1010200LTBL

 View more versions of this product

Catalog No. 18600230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Spectrum™ Chemical Acetic Acid, Glacial, Reagent, ACS , also known under the name ethanoic acid, it is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak Acid, when concentrated it is corrosive and can attack the skin. In its Glacial form (undiluted), it is used as a polar protic solvent. As a solvent it is used for purification by recrystallization of organic compounds and in the production of terephtalic acid. As an ACS grade Reagent, Spectrum Chemical manufactured Acetic Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications

Specifications

64-19-7
C2H4O2
UN2789
CC(O)=O
60.05
ACS
Drum
100%
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
99.7%
200 L
0.001%

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
CC(O)=O
C2H4O2
MFCD00036152
acetic acid
Videos
SDS
Documents

Documents

Promotions

Promotions