missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid, Glacial (USP/FCC/EP/BP), Fisher Chemical

Supplier:  Thermo Fisher Scientific A491212

Catalog No. A491-212


Explore available promotions

Description

Description

Glass Bottle; 2.5L

Specifications

Specifications

Acetic Acid, Glacial
17.4N
64-19-7
Colorless
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
99.0 to 100.5% (EP)
1.048g/cm³
40°C
Liquid
1.52 kPa at 20°C
2.5
>14.8°C
117°C
Clear and Colorless (EP/BP)
CH3COOH
2789
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
BP/EP/FCC/USP
2.5 L
Glass Bottle
0.01% max. (BP), 0.01% max. (EP), 0.01% max. (EP/BP)
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions