missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid, Optima™ LC/MS, Fisher Chemical™

Supplier:  Fisher Chemical A11350

 View more versions of this product

Catalog No. A11350


Explore available promotions

Description

Description

Specifications

Specifications

0.02 AU max. at 350nm, 0.05 AU max. at 280nm, 1 AU max. at 255nm
2.5
16°C
117°C
≥99.7 %
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
≥99.7%
1.048g/cm³
Nalgene™ Poly Bottle
5ppm max.
Acetic Acid
99.8%
64-19-7
Colorless
C2H4O2
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
OPTIMA™ LC-MS
50 mL
Liquid
1.52 kPa at 20°C

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0

SDS Documents

Documents

Promotions

Promotions