missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Acid Solution, 1N (Certified), Fisher Chemical

A volumetric standard solution, titrated at 25°C to pH 8.2 (phenolphthalein endpoint) against sodium hydroxide, which has been standardized against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific SA361

Catalog No. SA36-1



Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid Solution, 1N
Colorless
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
acetic acid
176
60.05
1N
Poly Bottle
64-19-7,7732-18-5
C2H4O2
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
0.995 to 1.005N
60.05
CHEBI:15366
Certified
1 L
Liquid

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

SDS Documents

Documents

Promotions

Promotions