missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic Anhydride (Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical A04004L06
Specifications
Acetic Anhydride | |
-73.1°C | |
1.087g/cm³ | |
140°C | |
≥97.0 % | |
C4H6O3 | |
MFCD00008705 | |
1715 | |
acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique | |
CC(=O)OC(=O)C | |
102.089 | |
CHEBI:36610 | |
≥97.0% | |
Liquid |
108-24-7 | |
Colorless | |
3 | |
53°C | |
Glass Bottle | |
(CH3CO)2O | |
4 L | |
0.002% max. | |
WFDIJRYMOXRFFG-UHFFFAOYSA-N | |
acetyl acetate | |
7918 | |
102.09 | |
Certified ACS |
Chemical Identifiers
108-24-7 | |
102.089 | |
WFDIJRYMOXRFFG-UHFFFAOYSA-N | |
7918 | |
acetyl acetate |
C4H6O3 | |
MFCD00008705 | |
acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique | |
CHEBI:36610 | |
CC(=O)OC(=O)C |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive