missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic Anhydride (Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical A0400500ML06

 View more versions of this product

Catalog No. 351001-500



Explore available promotions

Description

Description

Specifications

Specifications

Acetic Anhydride
-73.1°C
1.087g/cm³
140°C
≥97.0 %
C4H6O3
MFCD00008705
1715
acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
CC(=O)OC(=O)C
102.089
CHEBI:36610
≥97.0%
Liquid
108-24-7
Colorless
3
53°C
Glass Bottle
(CH3CO)2O
500 mL
0.002% max.
WFDIJRYMOXRFFG-UHFFFAOYSA-N
acetyl acetate
7918
102.09
Certified ACS

Chemical Identifiers

108-24-7
102.089
WFDIJRYMOXRFFG-UHFFFAOYSA-N
7918
acetyl acetate
C4H6O3
MFCD00008705
acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
CHEBI:36610
CC(=O)OC(=O)C
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions