missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetoacetamide, 97%, Thermo Scientific Chemicals

$71.28 - $71.28

Chemical Identifiers

CAS 5977-14-0
InChI Key GCPWJFKTWGFEHH-UHFFFAOYSA-N
Synonym acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
PubChem CID 80077
ChEBI CHEBI:28515
IUPAC Name 3-oxobutanamide
SMILES CC(=O)CC(=O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC442541000
SDS View Documents
Thermo Scientific Chemicals
442541000
100 g Glass bottle
Each for $71.28
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5977-14-0
acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
CHEBI:28515
CC(=O)CC(=O)N
GCPWJFKTWGFEHH-UHFFFAOYSA-N
80077
3-oxobutanamide

Specifications

5977-14-0
100.0
White to Yellow
Authentic
Glass bottle
CH3C(O)CH2C(O)NH2
acetoacetamide, acetylacetamide, acetoacetic acid amide, butanamide, 3-oxo, 3-oxobutanamid, unii-6o9u7cfy0t, 6o9u7cfy0t, 3-oxobutyramide, acmc-20alv9
GCPWJFKTWGFEHH-UHFFFAOYSA-N
3-oxobutanamide
80077
101.11
Crystalline Powder
96.0
-53°C to 56°C
113°C
96% min. (HPLC)
C4H7NO2
100 g
Solubility in water: miscible
CC(=O)CC(=O)N
101.11
CHEBI:28515
97%
Acetoacetamide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-774-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only