Learn More
Acetohydroxamic acid, 98+%
CAS: 546-88-3 | C2H5NO2 | 75.06 g/mol
$101.08 - $101.08
Chemical Identifiers
| CAS | 546-88-3 |
|---|---|
| Molecular Formula | C2H5NO2 |
| Molecular Weight (g/mol) | 75.06 |
| MDL Number | MFCD00009994 |
| InChI Key | RRUDCFGSUDOHDG-UHFFFAOYSA-N |
| Synonym | acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure |
| PubChem CID | 1990 |
| ChEBI | CHEBI:49029 |
| IUPAC Name | N-hydroxyacetamide |
| SMILES | CC(=O)NO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC170130050
|
Thermo Scientific Chemicals
170130050 |
5 g | Glass bottle |
Each for $101.08
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 546-88-3 | |
| 75.06 | |
| RRUDCFGSUDOHDG-UHFFFAOYSA-N | |
| 1990 | |
| N-hydroxyacetamide |
| C2H5NO2 | |
| MFCD00009994 | |
| acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure | |
| CHEBI:49029 | |
| CC(=O)NO |
Specifications
| 546-88-3 | |
| 100.0 | |
| White to Yellow | |
| 98+% | |
| C2H5NO2 | |
| MFCD00009994 | |
| 02, 187 | |
| 15, 62 | |
| Solubility in water: soluble | |
| CC(=O)NO | |
| 75.06 | |
| CHEBI:49029 | |
| 98+% | |
| Acetohydroxamic acid |
| 98.0 | |
| 86.0°C to 90.0°C | |
| Authentic | |
| Glass bottle | |
| CH3C(=NOH)OH | |
| 5 g | |
| 13,2 | |
| acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure | |
| RRUDCFGSUDOHDG-UHFFFAOYSA-N | |
| N-hydroxyacetamide | |
| 1990 | |
| 75.06 | |
| Crystalline Solid |
Safety and Handling
GHS H Statement:
May damage the unborn child.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-913-8
RUO – Research Use Only