Learn More
Acetoin, 96%, dimer
CAS: 23147-57-1 | C8H16O4 | 176.21 g/mol
$210.89 - $481.54
Chemical Identifiers
| CAS | 23147-57-1 |
|---|---|
| Molecular Formula | C8H16O4 |
| Molecular Weight (g/mol) | 176.21 |
| MDL Number | MFCD00038696 |
| InChI Key | DFMGATPNJMFDCR-UHFFFAOYNA-N |
| Synonym | acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl |
| PubChem CID | 179 |
| ChEBI | CHEBI:15688 |
| IUPAC Name | 3-hydroxybutan-2-one |
| SMILES | CC1OC(C)(O)C(C)OC1(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC411951000
|
Thermo Scientific Chemicals
411951000 |
100 g | Glass bottle |
Each for $210.89
|
|
||||
|
AC411955000
|
Thermo Scientific Chemicals
411955000 |
500 g | Glass bottle |
Each for $481.54
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 23147-57-1 | |
| 176.21 | |
| DFMGATPNJMFDCR-UHFFFAOYNA-N | |
| 179 | |
| 3-hydroxybutan-2-one |
| C8H16O4 | |
| MFCD00038696 | |
| acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl | |
| CHEBI:15688 | |
| CC1OC(C)(O)C(C)OC1(C)O |
Specifications
| 23147-57-1 | |
| 100.0 | |
| Green-Yellow to Yellow or White to Yellow | |
| 148.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4171 | |
| MFCD00038696 | |
| 01, 827 | |
| 15, 63 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol,propylene glycol,insoluble in vegetable oil | |
| CC1OC(C)(O)C(C)OC1(C)O | |
| 176.21 | |
| CHEBI:15688 | |
| 96% | |
| Acetoin, pract., 93% |
| 95.0 | |
| 15.0°C | |
| 1.0000g/mL | |
| 50°C | |
| 95% min. (GC) | |
| C8H16O4 | |
| CH3COCH(OH)CH3 | |
| 100 g | |
| 1 | |
| acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl | |
| DFMGATPNJMFDCR-UHFFFAOYNA-N | |
| 3-hydroxybutan-2-one | |
| 179 | |
| 88.11 | |
| Liquid (Monomer) or Powder or Crystals (Dimer) |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 208-174-1
RTECSNumber : EL8790000
TSCA : TSCA
Recommended Storage : may polymerize during storage
RUO – Research Use Only