missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetoin (May exist as crystalline dimer) 98.0+%, TCI America™

Supplier:  TCI America H022525G

 View more versions of this product

Catalog No. H022525G


Explore available promotions

Description

Description

Specifications

Specifications

Acetoin (May exist as crystalline dimer)
White-Yellow
C4H8O2
25 g
acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
CC(O)C(C)=O
88.11
CHEBI:15688
≥98.0% (GC)
513-86-0
144°C
MFCD00004521,MFCD00038696
2621
ROWKJAVDOGWPAT-UHFFFAOYNA-N
3-hydroxybutan-2-one
179
88.11
Crystalline Powder

Chemical Identifiers

513-86-0
88.11
ROWKJAVDOGWPAT-UHFFFAOYNA-N
179
3-hydroxybutan-2-one
C4H8O2
MFCD00004521,MFCD00038696
acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
CHEBI:15688
CC(O)C(C)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0597

RTECSNumber : EL8790000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions