missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, ACS Grade, ≥ 99.5%, LabChem™

Supplier:  LabChem LC104204

 View more versions of this product

Catalog No. LC104204


Explore available promotions

Description

Description

Specifications

Specifications

Acetone
Colorless
≤0.0003meq/gcSt
Carbon monoxide, Carbon dioxide
4 L
Amber Glass
≤0.001%
CH3COCH3
0.417mm2/s
≤0.5%
Soluble in water
CC(C)=O
58.08
CHEBI:15347
≥99.5%
67-64-1
100
≤0.0006meq/gcSt
786kg/m3
Passes Test
Liquid
C3H6O
MFCD00008765
UN1090
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
ACS

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness and dizziness.

GHS P Statement
Keep away from heat/hot surfaces, open flames.
No smoking.
Keep container tightly closed.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing mist/vapors/spray.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
If on skin (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
If inhaled: Remove person to fresh air and keep at rest in a posittion comfortable for breathing.
Call a poison center/doctor if you feel unwell.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container in accordance with local, state and federal regulations.

Danger

DOTInformation : DOT Class 3: Flammable Liquid

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions