missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetone, ACS Reagent, Reagents
Acetone, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C110250010E6
Specifications
Acetone, ACS Reagent | |
67-64-1 | |
56°C | |
Steel Pail | |
0.788 | |
MFCD00008765 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
ACS |
-94.7°C | |
Colorless | |
10 L | |
Liquid | |
C3H6O | |
Dimethyl Ketone, 2-Propanone | |
CC(C)=O | |
58.08 |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
Dimethyl Ketone, 2-Propanone | |
CC(C)=O |