missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone (Certified ACS), Fisher Chemical™

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Supplier:  Thermo Fisher Scientific Formerly Fairlawn BPA184

 View more versions of this product

Catalog No. A18-4

Explore available promotions

Description

Description

Specifications

Specifications

7
-95°C
Colorless
0.790g/cm³
4 L
Pass Test
Glass Bottle
0.001% max.
0.79
C3H6O
MFCD00008765
0.32 mPa/s at 20°C
0.5% max.
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(C)=O
58.08
CHEBI:15347
≥99.5%
Acetone
67-64-1
>4°C
56°C
-20°C
Conforms to reference
Liquid
Assay: ≥99.5%
0.0003mEq/g max.
0.0006mEq/g max.
247mbar at 20°C
1090
15, 65
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
Certified ACS Reagent

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions