missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone Dibutyl Acetal 95.0+%, TCI America™
Supplier: TCI America A078225ML
Specifications
| Acetone Dibutyl Acetal | |
| Yellow | |
| C11H24O2 | |
| 25 mL | |
| 2,2-Dibutoxypropane | |
| CCCCOC(C)(C)OCCCC | |
| 188.31 | |
| 188.31 | |
| Liquid |
| 141-72-0 | |
| 130°C | |
| MFCD00015248 | |
| 3271 | |
| CXBFTMAIVDLZLG-UHFFFAOYSA-N | |
| 1-[(2-butoxypropan-2-yl)oxy]butane | |
| 67335 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 141-72-0 | |
| 188.31 | |
| CXBFTMAIVDLZLG-UHFFFAOYSA-N | |
| 67335 | |
| CCCCOC(C)(C)OCCCC |
| C11H24O2 | |
| MFCD00015248 | |
| 2,2-Dibutoxypropane | |
| 1-[(2-butoxypropan-2-yl)oxy]butane |
Safety and Handling
TSCA : Yes