missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetone, For pesticide residue analysis, Honeywell Riedel-de Haën™
CHROMASOLV™, for pesticide residue analysis
$130.78 - $1007.05
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
6000210
|
Honeywell-Chromasolv
344801L |
1 L |
Each for $130.78
|
|
||||||
60046342
|
Honeywell Inc
344804X25L |
4 x 2.5 L |
Case of 4 Each for $978.08
|
|
||||||
6000212
|
Honeywell-Chromasolv
344804X4L |
4 x 4 L |
Case of 4 Each for $1,007.05
|
|
||||||
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
67-64-1 | |
0.791g/mL at 25°C | |
0.132 | |
C3H6O | |
MFCD00008765 | |
UN1090 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.8% (GC) | |
Acetone |
Colorless | |
1 L | |
−17°C (1.4°F) | |
CH3COCH3 | |
184 mmHg (20°C) | |
635680 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08g/mol | |
CHROMASOLV™ |
Safety and Handling
ShelfLife : 720 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone
EINECSNumber : 200-662-2
Recommended Storage : Room Temp