missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetone (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Supplier: Thermo Fisher Scientific Formerly Fairlawn BPA949SK4
Specifications
7 | |
-95°C | |
0.01 AU max. at 400nm, 0.02 AU max. at 350nm, 0.06 AU max. at 340nm, 1.00 AU max. at 330nm | |
>4°C | |
56°C | |
Pass Test | |
Liquid | |
1ppm max. | |
0.0003mEq/g max. | |
0.0006mEq/g max. | |
MFCD00008765 | |
0.32 mPa/s at 20°C | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.5% |
Acetone | |
67-64-1 | |
Colorless | |
0.790g/cm³ | |
4 L | |
Safe-Cote™ Amber Glass | |
Mobile phase for HPLC applications | |
99.5% min by GC | |
C3H6O | |
CH3COCH3 | |
247mbar at 20°C | |
0.5% max. | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 | |
HPLC |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open f
DOTInformation : DOT Class 3, : Flammable Liquid