missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone (NF/FCC), Fisher Chemical™

$295.08 - $10827.69

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A11-1
SDS View Documents
Thermo Fisher Scientific Formerly Fairlawn
BPA111
1 L Glass Bottle
Each for $295.08
Case of 6 Each for $1,770.48
 
A11S-4
SDS View Documents
Thermo Fisher Scientific Formerly Fairlawn
BPA11S4
4 L Aluminum Can
Each for $749.00
Case of 4 Each for $2,996.00
 
A11-4
SDS Accessories View Documents
Fisher Chemical
BPA114
4 L Glass Bottle
Each for $442.62
Case of 4 Each for $1,770.48
 
A11-20
SDS View Documents
Thermo Fisher Scientific Formerly Fairlawn
FLA1120
20 L Steel Pail
Each for $1,336.69
 
A11-200
SDS Accessories View Documents Promotion Details
Thermo Fisher Scientific Formerly Fairlawn
BPA11200
200 L Steel Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O

Specifications

7
67-64-1
10mg/kg max. (FCC)
0.790g/cm³
1 L
Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Liquid
0.789 Max. (NF), 0.7880 max. (FCC), 0.789 max. (NF)
CH3COCH3
247mbar at 20°C
1090; Colorless
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(C)=O
58.08
CHEBI:15347
NLT 99.5%, NMT 100.5% (FCC)
Acetone
-95°C
Colorless
>4°C
56°C
-20°C (-4°C)
Glass Bottle
1.358 to 1.360 (at 25°C) (FCC)
C3H6O
MFCD00008765
0.32 mPa/s at 20°C
0.5% Max. (FCC/NF), 0.5% max. (NF)
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
FCC/NF
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'