missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile with 0.05% Formic Acid (v/v), Fisher Chemical

Supplier:  Fisher Chemical HB98224

Catalog No. HB98224



Explore available promotions

Description

Description

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications

Specifications

Acetonitrile with 0.05% Formic Acid (v/v)
0.7810g/cm³
Pass Test
Liquid
C2H3N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
75-05-8,64-18-6
4 L
Glass Bottle
≥97.5 %
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
HPLC

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
Videos
SDS Documents

Documents

Promotions

Promotions