missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetonitrile with trifluoroacetic acid (0.05%), Mobile phase for HPLC, Fisher Chemical
Supplier: Fisher Chemical HB98124
Description
- Ideal mobile phase blend developed for use with HPLC instruments
- Pre-mixed for ease-of-use
- TFA is a common mobile phase additive that can function as an ion pair reagent
Specifications
Acetonitrile with 0.05% Trifluoroacetic Acid (v/v) | |
75-05-8,76-05-1 | |
4 L | |
Pass Test | |
Liquid | |
CH3CN | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
HPLC |
-45°C | |
81°C | |
6°C (43°F) | |
Glass Bottle | |
C2H3N | |
MFCD00001878 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
Inclusive between 0.0475% |
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Safety and Handling
Emergency Overview
Flammable liquid and vapor. Harmful by inhalation
SIGNAL WORD: DANGER!