missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™
0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, formisoton, formylic acid, collo-bueglatt, formira, aminic acid, bilorin, acide formique, myrmicyl, hydrogen carboxylic acid, methanoic acid, 2.5L
$294.88 - $1,206.69
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
LS120500
|
Thermo Fisher Scientific
LS120500 |
500mL | Glass Bottle |
N/A
|
|
||||
LS1201
|
Thermo Fisher Scientific
LS1201 |
1 L | Wide Neck Amber Bottle |
N/A
|
|
||||
LS120212
|
Thermo Fisher Scientific
LS120212 |
2.5 L | Glass Bottle |
N/A
|
|
||||
LS1204
|
Thermo Fisher Scientific
LS1204 |
4L | Glass Bottle |
N/A
|
|
||||
Description
0.1% Formic Acid in Acetonitrile (v/v), C2H3N, CAS Number-75-05-8, 64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 1L
- Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
- Pre-mixed for ease-of-use
- Low metal ion content to ensure minimal metal contamination
- Formic acid (FA) is a common mobile phase additive that provides protons for LC/MS analysis
- FA is also known to improve the peak shapes of the resulting separation
Chemical Identifiers
75-05-8 |
Specifications
LC-MS | |
500mL | |
Solvent Blends | |
Glass Bottle |
0.1% Formic Acid in Acetonitrile (v/v) | |
Liquid | |
75-05-8 | |
Pass Test |