missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mobile phase for HPLC, Acetonitrile with trifluoroacetic acid (0.1%), HPLC Grade, Fisher Chemical
Supplier: Thermo Fisher Scientific HB98134
Description
- Ideal mobile phase blend developed for use with HPLC instruments
- Pre-mixed for ease-of-use
- TFA is a common mobile phase additive that can function as an ion pair reagent
Specifications
2.0 | |
-50°C | |
>500°C | |
82°C | |
Pass Test | |
Liquid | |
C2H3N | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
Inclusive between 0.095% |
Acetonitrile with 0.1% Trifluoroacetic Acid (v/v) | |
75-05-8,76-05-1 | |
0.7810g/cm³ | |
4 L | |
Glass Bottle | |
Blended mobile phase for HPLC applications | |
MFCD00001878 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.04 | |
HPLC |
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |