missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, Anhydrous, 99.8% min, DriSolv, MilliporeSigma™

Supplier:  MilliporeSigma AX01436

Catalog No. MAX01436


Explore available promotions

Description

Description

Specifications

Specifications

Acetonitrile
1 L
99.8% min.
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
75-05-8
Liquid Free From Particulates
C2H3N
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05

Chemical Identifiers

75-05-8
41.053
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
C2H3N
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
Videos
SDS Documents

Documents

Promotions

Promotions