missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Honeywell Riedel-de Haën™

CHROMASOLV™, gradient grade, for HPLC,≥99.9%

$559.88 - $3358.03

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000242
View Documents
Honeywell-Chromasolv
348511L
1 L
Each for $559.88
 
6000243
View Documents
Honeywell-Chromasolv
3485125L
2.5 L
Each for $965.53
 
6000246
View Documents
Honeywell-Chromasolv
348516X1L
6 x 1 L
Case of 6 Each for $2,776.93
 
60046486
View Documents
Honeywell Chemicals
348514X25L
4 x 2.5 L
Case of 4 Each for $3,358.03
 
6000247
View Documents
Honeywell-Chromasolv
348514X4L
4 x 4 L
Case of 4 Each for $770.68
 
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

-48°C
HPLC-gradient/254nm ≤0.5mAU
81°C to 82°C
n20/D 1.344
CH3CN
72.8mmHg (20°C)
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.9%
Acetonitrile, for HPLC
75-05-8
0.786g/mL at 25°C
1 L
C2H3N
MFCD00001878
741857
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05g/mol
CHROMASOLV™ Gradient
Videos
Safety and Handling

Safety and Handling

ShelfLife : 720 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile

EINECSNumber : 200-835-2

Recommended Storage : Room Temp

SDS
missing translation for 'documents'

missing translation for 'documents'