missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile-d3, for NMR, 99.8 atom% D, AcroSeal™, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 217421000

Catalog No. AC217421000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
Specifications

Specifications

0.39 mPa.s (20°C)
Liquid
1.3430 to 1.3450
2°C
AcroSeal™ Glass Bottle
Colorless
2206-26-0
79.0°C to 81.0°C
CD3CN
02, IV, 428
WEVYAHXRMPXWCK-FIBGUPNXSA-N
2,2,2-trideuterioacetonitrile
123151
NMR
0.03% Max. (Coulometric)
0.8440g/mL
99.8% min. (NMR)
100 mL
Acetonitrile-d3
For NMR, 99.8 atom% D
-46.0°C
C2H3N
MFCD00001881
acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
[2H]C([2H])([2H])C#N
44.07
44.07

Chemical Identifiers

2206-26-0
44.07
WEVYAHXRMPXWCK-FIBGUPNXSA-N
123151
[2H]C([2H])([2H])C#N
C2H3N
MFCD00001881
acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
2,2,2-trideuterioacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only