missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile (HPLC), Fisher Chemical

HPLC mobile phase for analytical and preparative separations

$401.19 - $1451.42

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A998SK-1
SDS View Documents
Fisher Chemical
FLA998SK1
1 L Safe-Cote™ Amber Glass Bottle
Each for $578.67
Case of 6 Each for $3,472.02
 
A998-1
SDS View Documents
Fisher Chemical
FLA9981
1 L Wide Neck Amber Bottle
Each for $401.19
Case of 6 Each for $2,407.14
 
A998-4 Edge
SDS Accessories View Documents Promotion Details
Fisher Chemical
BPA9984
4 L Amber Glass Bottle
Each for $1,181.58
Case of 4 Each for $4,726.32
Edge
 
A998SK-4
SDS View Documents
Fisher Chemical
FLA998SK4
4 L Safe-Cote™ Amber Glass Bottle
Each for $1,451.42
Case of 4 Each for $5,805.68
 
Description

Description

Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

-46°C
0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test
>500°C
1 L
Pass Test
Liquid
Assay: 99.9% min by GC
0.008mEq/g max.
0.0006mEq/g max.
MFCD00001878
0.36 cP at 20°C
0.01% max.
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
Mobile phase for HPLC applications
75-05-8
Colorless
81°C
12.8°C (55°F)
Safe-Cote™ Amber Glass Bottle
1.0ppm max.
0.781
C2H3N
CH3CN
97mbar at 20°C
1648; Colorless
15, 69
Soluble in water
CC#N
41.053
CHEBI:38472
≥99.9%
Acetonitrile
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'