Learn More
Acetonitrile, HPLC Grade, ≥ 99.8%, LabChem™
$404.37 - $1144.27
Chemical Identifiers
| CAS | 75-05-8 |
|---|---|
| Molecular Formula | C2H3N |
| Molecular Weight (g/mol) | 41.053 |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
| PubChem CID | 6342 |
| ChEBI | CHEBI:38472 |
| IUPAC Name | acetonitrile |
| SMILES | CC#N |
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
| C2H3N | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
Specifications
| 75-05-8 | |
| Colorless | |
| ≤8μeq/g | |
| Carbon dioxide Carbon monoxide Nitrogen oxides cyanides | |
| 1 L | |
| Amber Glass | |
| ≤5ppm | |
| C2H3N | |
| 3.80mm2/s | |
| ≤0.1% | |
| Soluble in water | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| ≥99.8% | |
| Acetonitrile |
| Passes Test | |
| 100 | |
| ≤0.6μeq/g | |
| 787kg/m3 | |
| Passes Test | |
| Liquid | |
| 0.79g/mL | |
| CH3CN | |
| UN1648 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41.05 | |
| HPLC |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Toxic in contact with skin.
GHS P Statement
Keep away from heat/sparks/open flames/ hot surfaces.
No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Wear protective gloves, protective clothing, eye protection, face protection.
Store locked up in a well-ventilated place.
Keep cool.
Keep container tightly closed.
If on skin (or hair): Take off immediately all contaminated clothing.
Wash skin with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Wash contaminated clothing before reuse.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature