missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetonitrile, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™
$878.84 - $878.84
Chemical Identifiers
CAS | 75-05-08 |
---|---|
Molecular Weight (g/mol) | 41.05 |
Synonym | ACN, Methyl cyanide, Ethyl nitrile, Cyranomethane |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-100-233
|
Technidata
1000292500 |
2.5 L |
Each for $878.84
|
|
|||||
Chemical Identifiers
75-05-08 | |
41.05 | |
ACN, Methyl cyanide, Ethyl nitrile, Cyranomethane |
Specifications
-45.7°C | |
≤ 0.0001 meq/g | |
0.78 g/cm3 at 20 °C | |
2.5 L | |
2 °C | |
Glass Bottle | |
98.64 hPa at 20 °C | |
ACN, Methyl cyanide, Ethyl nitrile, Cyranomethane | |
≥99.97% | |
Acetonitrile |
75-05-08 | |
≤ 0.0002 meq/g | |
81.6°C | |
3 to 17% (V) | |
524 °C | |
≤ 1 mg/L | |
≤ 0.01% | |
41.05 | |
Hypergrade |