missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

Supplier:  Thermo Fisher Scientific A996SK4

 View more versions of this product

Catalog No. A996SK4

Explore available promotions

Description

Description

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 190-254nm range, to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only
Specifications

Specifications

Acetonitrile
75-05-8
Colorless
0.781g/cm³
4 L
Pass Test
Liquid
1.3405 to 1.3425 (at 25°C)
Assay: 99.9% min by GC
C2H3N
MFCD00001878
0.36 cP at 20°C
0.01% max.
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
OPTIMA™
-46°C
0.005 AU max. at 254nm, 0.02 AU max. at 220nm, 0.04 AU max. at 210nm, 0.05 AU max. at 205nm, 0.07 AU max. at 200nm, 0.15 AU max. at 195nm, 1.00 AU max. at 190nm
>500°C
81°C
49°C (120.2°F)
Safe-Cote™ Amber Glass
High purity mobile phase for HPLC and/or extraction solvent for GC applications
0.8ppm max.
0.008mEq/g max.
0.0006mEq/g max.
97mbar at 20°C
1648; Colorless
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.9%

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions