missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methyl cyanide, cyanomethane, acetonitrile, ethyl nitrile, ethanenitrile, cyanure de methyl, methanecarbonitrile, methylkyanid, methane, cyano, mecn, 2.5L

Supplier:  Fisher Chemical A955212

 View more versions of this product

Catalog No. A955212

Explore available promotions

Description

Description

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methane, cyano, ethyl nitrile, methanecarbonitrile, methylkyanid, mecn, acetonitrile, methyl cyanide, ethanenitrile, cyanure de methyl, cyanomethane, 1L

  • Ideal mobile phase for LC/MS and UHPLC-UV applications in pharmaceutical, biotechnology, clinical, environmental and food safety industries.
  • Novel gradient elution test with Photodiode Array Detector screens for UV-absorbing contaminants at every wavelength in the 200 to 400nm range to afford smooth baselines and to reduce interferences.
  • Evaluated for 17 metal impurities at ppb concentrations to minimize the formation of metal adducts during analysis and simplify spectral interpretation.
  • 0.1 micron filtration minimizes troublesome particles that may interfere with LC/MS and UHPLC system performance
  • Provides ideal solution to prepare cleaner, particle-free mobile phase blends. Positive mode ionization (as propazine) and negative mode ionization (as mecoprop) with ESI and Single Quadrupole MS detect organic contaminants at max 50ppb and max 300ppb, respectively
  • Glass bottles have fluorinated lining
Specifications

Specifications

Acetonitrile
75-05-8
Colorless
0.781g/cm³
2.5 L
Glass Bottle with fluorinated lining
Formulated for UHPLC-UV
Assay: 99.9% min by GC
≤0.008meq/g
≤0.0006meq/g
1878
0.36cp at 20°C
≤0.01%
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.9%
-46°C
≤1.00 AU at 190nm, ≤0.005 AU at 280nm, ≤0.15 AU at 195nm, ≤0.04 AU at 205nm, ≤0.04 AU at 205nm, ≤0.03 AU at 210nm, ≤0.025 AU at 215nm, ≤0.015 AU at 225nm, ≤0.015 AU at 225nm, ≤0.01 AU at 230nm, ≤0.005 AU at 254nm
>500°C
81°C
Pass Test
Liquid
≤0.8ppm
GC
C2H3N
CH3CN
97mbar at 20°C
1648
15, 69
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05
OPTIMA™ LC-MS

Chemical Identifiers

75-05-8
1878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health: 2
Flammability: 3
Instability: 0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions