missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetonyltriphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals
$111.22 - $370.71
Chemical Identifiers
CAS | 2236-01-3 |
---|---|
Molecular Formula | C21H20BrOP |
Molecular Weight (g/mol) | 399.27 |
MDL Number | MFCD00051863 |
InChI Key | KZBWHCFTAAHIJJ-UHFFFAOYSA-M |
Synonym | acetonyltriphenylphosphonium bromide, acetonyl triphenylphosphonium bromide, 2-oxopropyl triphenylphosphanium bromide, phosphonium, 2-oxopropyl triphenyl-, bromide, acetonyltriphenylphosphoniumbromide, acmc-1cmpe, 2-oxopropyl-triphenylphosphonium bromide, 2-oxopropyl triphenylphosphonium bromide, 2-oxopropyl triphenyl phosphanium bromide, phosphonium, 2-oxopropyl triphenyl-,bromide 1:1 |
PubChem CID | 638158 |
IUPAC Name | 2-oxopropyl(triphenyl)phosphanium;bromide |
SMILES | [Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2243406
|
Thermo Scientific Chemicals
B2243406 |
5 g |
Each for $111.22
|
|
|||||
AAB2243414
|
Thermo Scientific Chemicals
B2243414 |
25 g |
Each for $370.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2236-01-3 | |
399.27 | |
KZBWHCFTAAHIJJ-UHFFFAOYSA-M | |
638158 | |
[Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C21H20BrOP | |
MFCD00051863 | |
acetonyltriphenylphosphonium bromide, acetonyl triphenylphosphonium bromide, 2-oxopropyl triphenylphosphanium bromide, phosphonium, 2-oxopropyl triphenyl-, bromide, acetonyltriphenylphosphoniumbromide, acmc-1cmpe, 2-oxopropyl-triphenylphosphonium bromide, 2-oxopropyl triphenylphosphonium bromide, 2-oxopropyl triphenyl phosphanium bromide, phosphonium, 2-oxopropyl triphenyl-,bromide 1:1 | |
2-oxopropyl(triphenyl)phosphanium;bromide |
Specifications
2236-01-3 | |
C21H20BrOP | |
5 g | |
Hygroscopic | |
KZBWHCFTAAHIJJ-UHFFFAOYSA-M | |
2-oxopropyl(triphenyl)phosphanium;bromide | |
638158 | |
≥98% |
226°C to 227°C | |
MFCD00051863 | |
3802148 | |
acetonyltriphenylphosphonium bromide, acetonyl triphenylphosphonium bromide, 2-oxopropyl triphenylphosphanium bromide, phosphonium, 2-oxopropyl triphenyl-, bromide, acetonyltriphenylphosphoniumbromide, acmc-1cmpe, 2-oxopropyl-triphenylphosphonium bromide, 2-oxopropyl triphenylphosphonium bromide, 2-oxopropyl triphenyl phosphanium bromide, phosphonium, 2-oxopropyl triphenyl-,bromide 1:1 | |
[Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
399.27 | |
399.27 | |
Acetonyltriphenylphosphonium bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-796-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only