Learn More
Acetyl bromide, 98%, Thermo Scientific Chemicals
$137.61 - $368.11
Chemical Identifiers
CAS | 506-96-7 |
---|---|
Molecular Formula | C2H3BrO |
Molecular Weight (g/mol) | 122.95 |
MDL Number | MFCD00000114 |
InChI Key | FXXACINHVKSMDR-UHFFFAOYSA-N |
Synonym | ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide |
PubChem CID | 10482 |
IUPAC Name | acetyl bromide |
SMILES | CC(=O)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC159401000
|
Thermo Scientific Chemicals
159401000 |
100 mL |
Each for $137.61
|
|
|||||
AC159405000
|
Thermo Scientific Chemicals
159405000 |
500 mL |
Each for $368.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
506-96-7 | |
122.95 | |
FXXACINHVKSMDR-UHFFFAOYSA-N | |
10482 | |
CC(=O)Br |
C2H3BrO | |
MFCD00000114 | |
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
acetyl bromide |
Specifications
506-96-7 | |
100.0 | |
Colorless to Yellow | |
75°C to 77°C | |
Authentic | |
Glass bottle | |
1.4485 to 1.4505 | |
MFCD00000114 | |
02, 174 | |
1.66 | |
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
FXXACINHVKSMDR-UHFFFAOYSA-N | |
acetyl bromide | |
10482 | |
98% | |
Acetyl bromide, 98% |
97.5 | |
-96°C | |
1.6600g/mL | |
75°C | |
97.5% min. (Argentometry) | |
C2H3BrO | |
CH3COBr | |
100 mL | |
06,9; 13,208 | |
15, 77 | |
Solubility in water: reacts. Other solubilities: reacts with alcohol and acetone,soluble in ether,chloroform and benzene | |
CC(=O)Br | |
122.95 | |
122.95 | |
Fuming Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 208-061-7
RTECSNumber : AO5955000
TSCA : TSCA
Recommended Storage : May darken during storage
RUO – Research Use Only