missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl Bromide 97.0+%, TCI America™
Supplier: TCI America A0080500G
Specifications
Acetyl Bromide | |
-96°C | |
77°C | |
MFCD00000114 | |
1716 | |
FXXACINHVKSMDR-UHFFFAOYSA-N | |
acetyl bromide | |
10482 | |
≥97.0% (T) |
506-96-7 | |
Yellow | |
C2H3BrO | |
500 g | |
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
CC(=O)Br | |
122.949 | |
122.95 | |
Liquid |
Chemical Identifiers
506-96-7 | |
122.949 | |
FXXACINHVKSMDR-UHFFFAOYSA-N | |
10482 | |
CC(=O)Br |
C2H3BrO | |
MFCD00000114 | |
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
acetyl bromide |
Safety and Handling
EINECSNumber : (9)-0017
RTECSNumber : AO5955000
TSCA : Yes