missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl Chloride 98.0+%, TCI America™
$76.13 - $188.59
Chemical Identifiers
CAS | 75-36-5 |
---|---|
Molecular Formula | C2H3ClO |
Molecular Weight (g/mol) | 78.50 |
MDL Number | MFCD00000719 |
InChI Key | WETWJCDKMRHUPV-UHFFFAOYSA-N |
Synonym | ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 |
PubChem CID | 6367 |
ChEBI | CHEBI:37580 |
IUPAC Name | acetyl chloride |
SMILES | CC(Cl)=O |
Chemical Identifiers
75-36-5 | |
78.50 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
6367 | |
acetyl chloride |
C2H3ClO | |
MFCD00000719 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CHEBI:37580 | |
CC(Cl)=O |
Specifications
75-36-5 | |
Yellow | |
C2H3ClO | |
100 g | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
≥98.0% (GC,T) | |
Acetyl Chloride |
-112°C | |
52°C | |
MFCD00000719 | |
1717 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.50 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0631
RTECSNumber : AO6390000
TSCA : Yes
Recommended Storage : Refrigerator