missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210049005
Specifications
Acetyl Coenzyme A Trilithium Salt Trihydrate | |
White | |
C23H38Li3N7O17P3S | |
acetyl coenzyme a trilithium salt | |
[Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O | |
830.39 | |
881.1 |
75520-41-1 | |
>92% | |
5 mg | |
MOGXDJCKYPGZRO-JHJDYNLLSA-N | |
S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium | |
129893900 | |
Powder |
Chemical Identifiers
75520-41-1 | |
830.39 | |
acetyl coenzyme a trilithium salt | |
S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium |
C23H38Li3N7O17P3S | |
MOGXDJCKYPGZRO-JHJDYNLLSA-N | |
129893900 | |
[Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O |
Safety and Handling
Recommended Storage : Store at -20°C.