missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210049025

 View more versions of this product

Catalog No. ICN10049025


Explore available promotions

Description

Description

Specifications

Specifications

Acetyl Coenzyme A Trilithium Salt Trihydrate
White
C23H38Li3N7O17P3S
acetyl coenzyme a trilithium salt
[Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O
830.39
881.1
75520-41-1
>92%
25 mg
MOGXDJCKYPGZRO-JHJDYNLLSA-N
S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium
129893900
Powder

Chemical Identifiers

75520-41-1
830.39
acetyl coenzyme a trilithium salt
S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium
C23H38Li3N7O17P3S
MOGXDJCKYPGZRO-JHJDYNLLSA-N
129893900
[Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C.

SDS
Documents

Documents

Promotions

Promotions