missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylcholine Bromide 98.0+%, TCI America™

Supplier:  TCI America A008325G

Catalog No. A008325G

Explore available promotions

Description

Description

Specifications

Specifications

Acetylcholine Bromide
146°C
C7H16BrNO2
25 g
ZEHGKSPCAMLJDC-UHFFFAOYSA-M
2-acetyloxyethyl(trimethyl)azanium;bromide
65551
226.11
Crystalline Powder
66-23-9
White
MFCD00011814
acetylcholine bromide, pragmoline, tonocholin b, acetylcholine bromhydrate, acetylcholine hydrobromide, 2-acetoxy-n,n,n-trimethylethanaminium bromide, trimethyl 2-acetoxyethyl ammonium bromide, acetoxyethyl-trimethylammonium bromide, 2-acetoxyethyl trimethylammonium bromide, choline, acetyl-, bromide
CC(=O)OCC[N+](C)(C)C.[Br-]
226.114
CHEBI:55316
≥98.0% (T)

Chemical Identifiers

66-23-9
226.114
ZEHGKSPCAMLJDC-UHFFFAOYSA-M
65551
2-acetyloxyethyl(trimethyl)azanium;bromide
C7H16BrNO2
MFCD00011814
acetylcholine bromide, pragmoline, tonocholin b, acetylcholine bromhydrate, acetylcholine hydrobromide, 2-acetoxy-n,n,n-trimethylethanaminium bromide, trimethyl 2-acetoxyethyl ammonium bromide, acetoxyethyl-trimethylammonium bromide, 2-acetoxyethyl trimethylammonium bromide, choline, acetyl-, bromide
CHEBI:55316
CC(=O)OCC[N+](C)(C)C.[Br-]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0249&(9)-1504

RTECSNumber : FZ9680000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions