missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine Chloride, 99%

CAS: 60-31-1 | C7H16ClNO2 | 181.66 g/mol

Supplier:  Thermo Scientific Chemicals 159171000

 View more versions of this product

Catalog No. AC159171000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetylcholine chloride
98.5
149.0°C to 152.0°C
1.0% max. (105°C, 3 hrs)
98.5% min. (on dried basis) (Argentometry)
C7H16ClNO2
MFCD00011698
04,III,655
acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
JUGOREOARAHOCO-UHFFFAOYSA-M
2-acetyloxyethyl(trimethyl)azanium;chloride
6060
181.66
Crystalline Powder
60-31-1
100.0
White
Authentic
Glass bottle
CH3CO2CH2CH2N(CH3)3Cl
100 g
15, 82
Solubility in water: soluble in cold water. Other solubilities: decomposes by hot water or alkalies, soluble in alcohol, chloroform (18.2g/L at 23 °C), practically insoluble in ether
CC(=O)OCC[N+](C)(C)C.[Cl-]
181.66
CHEBI:2417
99%

Chemical Identifiers

60-31-1
181.66
JUGOREOARAHOCO-UHFFFAOYSA-M
6060
2-acetyloxyethyl(trimethyl)azanium;chloride
C7H16ClNO2
MFCD00011698
acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
CHEBI:2417
CC(=O)OCC[N+](C)(C)C.[Cl-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-468-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only