missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetylcholine iodide, 98%, Thermo Scientific Chemicals

$668.38 - $668.38

Chemical Identifiers

CAS 2260-50-6
Molecular Formula C7H16INO2
Molecular Weight (g/mol) 273.11
MDL Number MFCD00011815
InChI Key SMBBQHHYSLHDHF-UHFFFAOYSA-M
Synonym acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
PubChem CID 75271
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;iodide
SMILES [I-].CC(=O)OCC[N+](C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1169414
View Documents
Thermo Scientific Chemicals
A1169414
25 g
Each for $668.38
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2260-50-6
273.11
SMBBQHHYSLHDHF-UHFFFAOYSA-M
75271
[I-].CC(=O)OCC[N+](C)(C)C
C7H16INO2
MFCD00011815
acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
2-acetyloxyethyl(trimethyl)azanium;iodide

Specifications

2260-50-6
C7H16INO2
25 g
Hygroscopic; Light sensitive
SMBBQHHYSLHDHF-UHFFFAOYSA-M
2-acetyloxyethyl(trimethyl)azanium;iodide
75271
98%
161°C to 165°C
MFCD00011815
3571339
acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
[I-].CC(=O)OCC[N+](C)(C)C
273.11
273.11
Acetylcholine iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 218-862-3

RTECSNumber : KH3300000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only