missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acid Blue 29, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215259680
Description
Stains and dyesSpecifications
Acid Blue 29 | |
C22H14N6Na2O9S2 | |
C.I. 20460 | |
[Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O | |
616.49 | |
616.5 | |
100 g |
5850-35-1 | |
MFCD00009960 | |
WEILTBATXPPXPP-AXSRUCDFSA-L | |
disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate | |
44134491 | |
Black | |
Powder |
Chemical Identifiers
5850-35-1 | |
616.49 | |
WEILTBATXPPXPP-AXSRUCDFSA-L | |
44134491 | |
[Na+].[Na+].NC1=C(N=NC2=CC=CC(=C2)[N+]([O-])=O)C(=CC2=C1C(=O)\C(=N/NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O |
C22H14N6Na2O9S2 | |
MFCD00009960 | |
C.I. 20460 | |
disodium (3E)-5-amino-6-[2-(3-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate |
Safety and Handling
EINECSNumber : 227-449-7
Recommended Storage : Store at Room Temperature (15° to 30°C)