missing translation for 'onlineSavingsMsg'
Learn More

Acid Green 1 80.0+%, TCI America™

$89.48 - $838.83

Chemical Identifiers

CAS 19381-50-1
Molecular Formula C30H18FeN3Na3O15S3
Molecular Weight (g/mol) 881.474
MDL Number MFCD00003886
InChI Key VLBLPLXOYXEXJK-UHFFFAOYSA-K
Synonym Naphthol Green B
PubChem CID 14598749
IUPAC Name trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron
SMILES C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N030625G
View Documents
TCI America
N030625G
25 g
Each for $89.48
Only null left
 
N0306500G
View Documents
TCI America
N0306500G
500 g
Each for $838.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

19381-50-1
881.474
VLBLPLXOYXEXJK-UHFFFAOYSA-K
14598749
C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe]
C30H18FeN3Na3O15S3
MFCD00003886
Naphthol Green B
trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron

Specifications

19381-50-1
MFCD00003886
VLBLPLXOYXEXJK-UHFFFAOYSA-K
trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron
14598749
≥80.0% (E)
25 g
Acid Green 1
C30H18FeN3Na3O15S3
Naphthol Green B
C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe]
881.474
878.45
Black/Green
Crystal/Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-4373

RTECSNumber : LJ8930000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'