missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acrylamide: Bis-Acrylamide 19:1 (40% Solution/Electrophoresis), Fisher BioReagents
Supplier: Fisher BioReagents BP14061
Description
This Acrylamide Solution is premixed for 5% cross-linking and is suitable for DNA sequencing and separation of low-molecular-weight proteins.Specifications
Acrylamide: Bis-Acrylamide 19:1 | |
Liquid | |
10μmhos max. | |
1 L | |
DNase free | |
MFCD00008032 | |
NC(=O)C=C | |
71.08 | |
CHEBI:28619 | |
Polypropylene Bottle |
40% Solution | |
79-06-1,110-26-9,7732-18-5 | |
Colorless | |
DNase-, RNase- and Protease-Free | |
C3H5NO | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
Electrophoresis |
Chemical Identifiers
79-06-1 | |
71.08 | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
CHEBI:28619 | |
NC(=O)C=C |
C3H5NO | |
MFCD00008032 | |
6579 | |
prop-2-enamide |
Safety and Handling
WARNING!
Emergency Overview
Suspect cancer hazard. May cause heritable genetic damage. Toxic if swallowed. Harmful if absorbed through skin or if inhaled. Irritating to eyes and skin. May cause irritation of respiratory tract. May cause an allergic skin reaction. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Hazardous polymerization may occur. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood.
NFPA
Health:2
Flammability:2
Instability:2
Recommended Storage : 4°C