missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acrylamide: Bis-Acrylamide 29:1 (40% Solution/Electrophoresis), Fisher BioReagents
Supplier: Fisher BioReagents BP14081
Description
PP Bottle; 1 L
This Acrylamide Solution is premixed for 3.3% cross-linking and is suitable for DNA sequencing and separation of proteins.Specifications
Acrylamide: Bis-Acrylamide 29:1 | |
Liquid | |
10μmhos max. | |
Colorless | |
Polypropylene Bottle | |
C3H5NO | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
Electrophoresis |
40% Solution | |
79-06-1,110-26-9,7732-18-5 | |
1.035 g/mL | |
1 L | |
DNase-, RNase- and Protease-Free | |
MFCD00008032 | |
NC(=O)C=C | |
71.08 | |
CHEBI:28619 |