missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acrylamide: Bis-Acrylamide 37.5:1 (40% Solution/Electrophoresis), Fisher BioReagents

Supplier:  Fisher BioReagents BP14101

Catalog No. BP1410-1


Explore available promotions

Description

Description

This Acrylamide Solution is premixed for 2.6% cross-linking and is suitable for separation of high-molecular-weight proteins.
Specifications

Specifications

Acrylamide: Bis-Acrylamide 37.5:1
Liquid
10μmhos max.
1 L
C3H5NO
HRPVXLWXLXDGHG-UHFFFAOYSA-N
prop-2-enamide
6579
Electrophoresis
40% Solution
79-06-1,110-26-9,7732-18-5
Colorless
DNase-, RNase- and Protease-Free
MFCD00008032
NC(=O)C=C
71.08
CHEBI:28619
Polypropylene Bottle

Chemical Identifiers

79-06-1
71.08
HRPVXLWXLXDGHG-UHFFFAOYSA-N
CHEBI:28619
NC(=O)C=C
C3H5NO
MFCD00008032
6579
prop-2-enamide
Videos
Safety and Handling

Safety and Handling

Recommended Storage : 4°C

SDS Documents

Documents

Promotions

Promotions