Learn More
Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
Supplier: Fisher BioReagents BP1705
Description
This Acrylamide is 3X recrystallized to obtain a high-purity crystalline material suitable for electrophoretic separation of proteins and nucleic acids.Specifications
Acrylamide | |
0.002% max. | |
125°C | |
2.50μmhos max. | |
6.5 to 8.0 | |
DNase free | |
DNase-, RNase- and Protease-Free | |
C3H5NO | |
MFCD00008032 | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
71.08 | |
Electrophoresis |
Solid | |
79-06-1 | |
82°C | |
White | |
5 kg | |
Poly Pail | |
≥99.9 % | |
CH2:CHCONH2 | |
15, 120 | |
NC(=O)C=C | |
71.08 | |
CHEBI:28619 | |
≥99.9% |
Chemical Identifiers
79-06-1 | |
71.08 | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
CHEBI:28619 | |
NC(=O)C=C |
C3H5NO | |
MFCD00008032 | |
6579 | |
prop-2-enamide |
Safety and Handling
WARNING!
Emergency Overview
Toxic if swallowed. May cause cancer. Harmful if absorbed through skin or if inhaled. May impair fertility. May cause heritable genetic damage. May cause an allergic skin reaction. Irritating to eyes and skin. Toxic: danger of serious damage to health by prolonged exposure through inhalation, in contact with. May cause central nervous system effects. Do not breathe dust. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Do not get in eyes, on skin, or on clothing. Keep away from heat, sparks and open flame - No smoking.
NFPA
Health:2
Flammability:2
Instability:2
DOTInformation : DOT Class 6.1, : Poison
Recommended Storage : RT