missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents

Supplier:  Fisher BioReagents BP170500

 View more versions of this product

Catalog No. BP170-500



Explore available promotions

Description

Description

This Acrylamide is 3X recrystallized to obtain a high-purity crystalline material suitable for electrophoretic separation of proteins and nucleic acids.
Specifications

Specifications

Acrylamide
0.002% max.
125°C
2.50μmhos max.
6.5 to 8.0
DNase free
DNase-, RNase- and Protease-Free
C3H5NO
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
prop-2-enamide
6579
71.08
Electrophoresis
Solid
79-06-1
82°C
White
500 g
Poly Bottle
≥99.9 %
CH2:CHCONH2
15, 120
NC(=O)C=C
71.08
CHEBI:28619
≥99.9%

Chemical Identifiers

79-06-1
71.08
HRPVXLWXLXDGHG-UHFFFAOYSA-N
CHEBI:28619
NC(=O)C=C
C3H5NO
MFCD00008032
6579
prop-2-enamide
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Toxic if swallowed. May cause cancer. Harmful if absorbed through skin or if inhaled. May impair fertility. May cause heritable genetic damage. May cause an allergic skin reaction. Irritating to eyes and skin. Toxic: danger of serious damage to health by prolonged exposure through inhalation, in contact with. May cause central nervous system effects. Do not breathe dust. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Do not get in eyes, on skin, or on clothing. Keep away from heat, sparks and open flame - No smoking.

NFPA

Health:2
Flammability:2
Instability:2

DOTInformation : DOT Class 6.1, : Poison

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions