missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acrylic acid, low water content, 99.5%, stab. with ca 200ppm 4-methoxyphenol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 043359K4

 View more versions of this product

Catalog No. AA43359K4


Explore available promotions

Description

Description

Acrylic acid acts as a precursor in the production of 3-hydroxypropionic acid. It is used in the preparation of water-absorbent resins. It reacts with alcohols to prepare the corresponding esters. Its esters are used as raw materials for synthetic resins, rubbers, coating adhesives, water-based paints, floor polishes and adhesives. It is also used to prepare forming homopolymers or copolymers by reacting with other monomers like acrylamides, acrylonitrile, vinyl, styrene, and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acrylic acid
79-10-7
1.051 g/mL
54°C (130°F)
C3H4O2
MFCD00004367
UN2218
14,130
Miscible with water.
C=CC(=O)O
72.063
CHEBI:18308
99.5%
Low water content, Stabilized with ≈200ppm 4-methoxyphenol
∼13°C
138°C to 139°C
Pungent
1.4202
2.5 L
635743
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
NIXOWILDQLNWCW-UHFFFAOYSA-N
prop-2-enoic acid
6581
72.06
Liquid

Chemical Identifiers

79-10-7
72.063
NIXOWILDQLNWCW-UHFFFAOYSA-N
6581
prop-2-enoic acid
C3H4O2
MFCD00004367
acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomer
CHEBI:18308
C=CC(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACRYLIC ACID, STABILIZED

EINECSNumber : 201-177-9

RTECSNumber : AS4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only