missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Acrylic Burette with PTFE Key Stopcock, Class B

Transparent acrylic burette features easy to read graduations and leakproof PTFE plunger stopcock.

$57.29 - $71.71

Specifications

Autoclavable Not Autoclavable
Color Clear
Material Acrylic
Graduations 0.10 mL
For Use With (Application) Used in quantitative chemical analysis to measure the volume of a liquid or a gas.
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S27028
View Documents
Eisco™
CH0233A
25 mL
Each for $71.71
 
S95180A
View Documents
Eisco™
CH0233B
50 mL
Each for $57.29
 
Description

Description

  • Made of high quality acrylic, highly resistant to impact and shattering
  • Acrylic body is resistant to dilute minerals, acids and bases except hydrofluoric acid and ammonium hydroxide
  • Includes interchangeable, nonstick, chemical resistant PTFE key stopcock
  • Calibrated to deliver
  • Clean with non-abrasive cleaners, including strong detergents
  • Do not autoclave
  • Replacement stopcocks available; S24498
Specifications

Specifications

Not Autoclavable
Acrylic
Used in quantitative chemical analysis to measure the volume of a liquid or a gas.
Key
Buret
Clear
0.10 mL
Blue print scale
PTFE
Class B
SDS
missing translation for 'documents'

missing translation for 'documents'