United Scientific™ Acrylic Demonstration Lens Set DFS Item
Set of six lenses of a variety of types.
Supplier:  United Scientific™ LSTA75
 View more versions of this product
Catalog No. S42923
Explore available promotions
Description
Description
  • Each single-diameter set includes a double convex, plano-convex, concavo-convex, double concave, plano-concave and convexo-concave lens
  • Three diameters available
  • Includes an activity guide
Specifications
Specifications
Lens Set
6 Lenses, one each of double convex, plano convex, concavo convex, double concave, plano concave, and convexo concave lens, activity guide and a reusable cardboard storage box
3 in.
Teaching and demonstrating the basic principles of optics
6 Pk.
75 mm