missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin, from Rabbit Muscle, Thermo Scientific Chemicals
Description
Used in cell transport studies, and inhibits deoxyribonuclease I
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| For Use With (Application) | Cell Transport |
| Formulation | Lyophilized powder |
| Quantity | 1mg |
| Source | Rabbit muscle |
| Storage Requirements | -30° to -10°C |
| Regulatory Status | RUO |
| Product Type | Actin, from Rabbit Muscle |
| Recombinant | Native |
| Form | Lyophilized |
RUO – Research Use Only