missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Actinomycin D, >99%, MP Biomedicals™
$71.60 - $2631.49
Chemical Identifiers
CAS | 50-76-0 |
---|---|
Molecular Formula | C62H86N12O16 |
Molecular Weight (g/mol) | 1255.438 |
InChI Key | RJURFGZVJUQBHK-HUZQGMAJSA-N |
Synonym | Dactinomycin, Actinomycin-[thr-val-pro-sar-meval] |
PubChem CID | 131954673 |
IUPAC Name | 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, |
SMILES | CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10465801
|
MP Biomedicals Inc
0210465801 |
1 mg |
Each for $71.60
|
|
|||||
ICN10465810
|
MP Biomedicals Inc
0210465810 |
10 mg |
Each for $309.77
|
|
|||||
ICN10465850
|
MP Biomedicals Inc
0210465850 |
50 mg |
Each for $1,341.96
|
|
|||||
ICN10465880
|
MP Biomedicals Inc
0210465880 |
100 mg |
Each for $2,631.49
|
|
|||||
Chemical Identifiers
50-76-0 | |
1255.438 | |
Dactinomycin, Actinomycin-[thr-val-pro-sar-meval] | |
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, |
C62H86N12O16 | |
RJURFGZVJUQBHK-HUZQGMAJSA-N | |
131954673 | |
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C |
Specifications
Actinomycin D | |
50-76-0 | |
>99% | |
Dactinomycin, Actinomycin-[thr-val-pro-sar-meval] | |
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C | |
1255.438 | |
1255.5 |
Powder | |
1 mg | |
C62H86N12O16 | |
RJURFGZVJUQBHK-HUZQGMAJSA-N | |
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4, | |
131954673 | |
Actinomycin D |